Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCCTTTCTCACGCAAAGCCATT[C/T]CCACAAATCCAGACCATACCATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606564 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PMEPA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PMEPA1 - prostate transmembrane protein, androgen induced 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001255976.1 | 3787 | UTR 3 | NP_001242905.1 | |||
NM_020182.4 | 3787 | UTR 3 | NP_064567.2 | |||
NM_199169.2 | 3787 | UTR 3 | NP_954638.1 | |||
NM_199170.2 | 3787 | UTR 3 | NP_954639.1 | |||
NM_199171.2 | 3787 | UTR 3 | NP_954640.1 |