Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTCTCTTCCTAGACAATAAGCT[A/C]ATGGTGTTTATAACAGAACAACATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605114 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4325 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR4325 - microRNA 4325 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPO11 - SPO11, initiator of meiotic double stranded breaks | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012444.2 | Intron | NP_036576.1 | ||||
NM_198265.1 | Intron | NP_937998.1 | ||||
XM_005260379.3 | Intron | XP_005260436.1 | ||||
XM_005260380.3 | Intron | XP_005260437.1 | ||||
XM_005260381.3 | Intron | XP_005260438.1 | ||||
XM_005260382.4 | Intron | XP_005260439.1 | ||||
XM_011528756.2 | Intron | XP_011527058.1 | ||||
XM_011528757.2 | Intron | XP_011527059.1 |
Set Membership: |
HapMap |