Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATATAGACATAGAAAAGCTTTGA[C/T]GGAAAGTACCCTCATTTATTATATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603566 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALML4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CALML4 - calmodulin like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031733.2 | 3109 | UTR 3 | NP_001026903.2 | |||
NM_001286694.1 | 3109 | UTR 3 | NP_001273623.1 | |||
NM_001286695.1 | 3109 | UTR 3 | NP_001273624.1 | |||
NM_033429.2 | 3109 | UTR 3 | NP_219501.2 |
PIAS1 - protein inhibitor of activated STAT 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320687.1 | 3109 | Intron | NP_001307616.1 | |||
NM_016166.2 | 3109 | Intron | NP_057250.1 | |||
XM_011522126.2 | 3109 | Intron | XP_011520428.1 | |||
XM_011522127.2 | 3109 | Intron | XP_011520429.1 | |||
XM_017022688.1 | 3109 | Intron | XP_016878177.1 | |||
XM_017022689.1 | 3109 | Intron | XP_016878178.1 | |||
XM_017022690.1 | 3109 | Intron | XP_016878179.1 | |||
XM_017022691.1 | 3109 | Intron | XP_016878180.1 |
Set Membership: |
HapMap |