Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTAAAGAAATTCCTGTGAAGAG[C/T]CCCTTTGTAACACAAATCTTATTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612538 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC49 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC49 - leucine rich repeat containing 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199017.2 | Intron | NP_001185946.1 | ||||
NM_001199018.2 | Intron | NP_001185947.1 | ||||
NM_001284357.1 | Intron | NP_001271286.1 | ||||
NM_017691.4 | Intron | NP_060161.2 | ||||
XM_005254492.4 | Intron | XP_005254549.1 | ||||
XM_011521715.2 | Intron | XP_011520017.1 | ||||
XM_011521717.2 | Intron | XP_011520019.1 | ||||
XM_011521718.2 | Intron | XP_011520020.1 | ||||
XM_017022357.1 | Intron | XP_016877846.1 | ||||
XM_017022358.1 | Intron | XP_016877847.1 | ||||
XM_017022359.1 | Intron | XP_016877848.1 |
THAP10 - THAP domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020147.3 | Intron | NP_064532.1 |