Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGTGAATGAGTAGGCATAAGA[C/T]TGGCACTTTCTCTTAATGTTGGAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 610005 | |||||||||||||||||||||||
Literature Links: |
TNIK PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TNIK - TRAF2 and NCK interacting kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161560.2 | Intron | NP_001155032.1 | ||||
NM_001161561.2 | Intron | NP_001155033.1 | ||||
NM_001161562.2 | Intron | NP_001155034.1 | ||||
NM_001161563.2 | Intron | NP_001155035.1 | ||||
NM_001161564.2 | Intron | NP_001155036.1 | ||||
NM_001161565.2 | Intron | NP_001155037.1 | ||||
NM_001161566.2 | Intron | NP_001155038.1 | ||||
NM_015028.3 | Intron | NP_055843.1 |