Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATAAAAAGGAGAGTAATAGGAGA[A/G]TTTGGAGTGGATGGAAAAGAAAGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611622 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQCJ PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IQCJ - IQ motif containing J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042705.2 | Intron | NP_001036170.1 | ||||
NM_001042706.2 | Intron | NP_001036171.1 | ||||
NM_001197100.1 | Intron | NP_001184029.1 |
IQCJ-SCHIP1 - IQCJ-SCHIP1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197113.1 | Intron | NP_001184042.1 | ||||
NM_001197114.1 | Intron | NP_001184043.1 |