Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGTTCGTTTAAAAGTTGTGCTAC[G/T]TTTGAGGTGAAGTTAAGAGTGTCTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 106165 | |||||||||||||||||||||||
Literature Links: |
AGTR1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
AGTR1 - angiotensin II receptor type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000685.4 | Intron | NP_000676.1 | ||||
NM_004835.4 | Intron | NP_004826.5 | ||||
NM_009585.3 | Intron | NP_033611.1 | ||||
NM_031850.3 | Intron | NP_114038.4 | ||||
NM_032049.3 | Intron | NP_114438.2 |