Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTTATTGAAGCAATAGTTTTGT[C/T]CTCTACATCAATTCTGATTTCAGTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616149 | |||||||||||||||||||||||
Literature Links: |
SLC25A36 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLC25A36 - solute carrier family 25 member 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104647.1 | Intron | NP_001098117.1 | ||||
NM_018155.2 | Intron | NP_060625.2 | ||||
XM_005247575.4 | Intron | XP_005247632.1 |