Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCCTTTACCTGCCACTGTGGGT[A/T]TTGTCCAAGGAAGCAGGTATGAGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602512 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RGS12 - regulator of G-protein signaling 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002926.3 | Intron | NP_002917.1 | ||||
NM_198227.1 | Intron | NP_937870.1 | ||||
NM_198229.2 | Intron | NP_937872.1 | ||||
XM_006713905.1 | Intron | XP_006713968.1 | ||||
XM_006713906.2 | Intron | XP_006713969.1 | ||||
XM_006713907.1 | Intron | XP_006713970.1 | ||||
XM_011513543.2 | Intron | XP_011511845.1 | ||||
XM_017008529.1 | Intron | XP_016864018.1 | ||||
XM_017008530.1 | Intron | XP_016864019.1 | ||||
XM_017008531.1 | Intron | XP_016864020.1 | ||||
XM_017008532.1 | Intron | XP_016864021.1 | ||||
XM_017008533.1 | Intron | XP_016864022.1 | ||||
XM_017008534.1 | Intron | XP_016864023.1 |