Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGGCTGCTGGGACCCAGTCCCA[C/G]GCCTCCAGCCACCAGGCACCATAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610669 | ||||||||||||||||||||
Literature Links: |
FAM193A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM193A - family with sequence similarity 193 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNIP2 - TNFAIP3 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161527.1 | 1217 | Missense Mutation | CCT,CGT | P,R 270 | NP_001154999.1 | |
NM_001292016.1 | 1217 | Missense Mutation | CCT,CGT | P,R 294 | NP_001278945.1 | |
NM_024309.3 | 1217 | Missense Mutation | CCT,CGT | P,R 377 | NP_077285.3 |