Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAGATTTGCACATTTCAGTGT[A/G]TGTAAATTCGACTCCTAAAAACTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616585 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM192A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM192A - family with sequence similarity 192 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RSPRY1 - ring finger and SPRY domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305163.1 | Intron | NP_001292092.1 | ||||
NM_001305164.1 | Intron | NP_001292093.1 | ||||
NM_001305182.1 | Intron | NP_001292111.1 | ||||
NM_133368.2 | Intron | NP_588609.1 | ||||
XM_005256220.1 | Intron | XP_005256277.1 | ||||
XM_011523427.1 | Intron | XP_011521729.1 | ||||
XM_011523428.1 | Intron | XP_011521730.1 | ||||
XM_011523430.1 | Intron | XP_011521732.1 | ||||
XM_017023844.1 | Intron | XP_016879333.1 |