Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACGTCCCACTGCCTTCTGAGAG[C/G]GCTATAAAGGCAGCGGAAGGGTAGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605646 | |||||||||||||||||||||||
Literature Links: |
SLC26A4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLC26A4 - solute carrier family 26 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000441.1 | 159 | UTR 5 | NP_000432.1 | |||
XM_005250425.2 | 159 | Intron | XP_005250482.1 | |||
XM_006716025.3 | 159 | Intron | XP_006716088.1 | |||
XM_017012318.1 | 159 | Intron | XP_016867807.1 |
SLC26A4-AS1 - SLC26A4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |