Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTCTCCGTGGCATAGTTCCCAA[G/T]AGACTCCTATGTATCGAAAGCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118493 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHRM2 - cholinergic receptor muscarinic 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000739.2 | Intron | NP_000730.1 | ||||
NM_001006626.1 | Intron | NP_001006627.1 | ||||
NM_001006627.1 | Intron | NP_001006628.1 | ||||
NM_001006628.1 | Intron | NP_001006629.1 | ||||
NM_001006629.1 | Intron | NP_001006630.1 | ||||
NM_001006630.1 | Intron | NP_001006631.1 | ||||
NM_001006631.1 | Intron | NP_001006632.1 | ||||
NM_001006632.1 | Intron | NP_001006633.1 | ||||
XM_011515769.2 | Intron | XP_011514071.1 |
Set Membership: |
HapMap |