Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAAGTTCACAGAGGTGGGGTCTC[C/T]AGACAGGAGTAAGGCACACGGTGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176941 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TYK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TYK2 - tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003331.4 | 3811 | UTR 3 | NP_003322.3 | |||
XM_011528245.1 | 3811 | UTR 3 | XP_011526547.1 | |||
XM_011528246.2 | 3811 | UTR 3 | XP_011526548.1 | |||
XM_011528247.1 | 3811 | UTR 3 | XP_011526549.1 | |||
XM_011528249.2 | 3811 | UTR 3 | XP_011526551.1 | |||
XM_011528251.1 | 3811 | UTR 3 | XP_011526553.1 | |||
XM_011528252.1 | 3811 | Intron | XP_011526554.1 |