Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCCCTGTCACCCCAGCCTTCC[A/G]CAGGGTCAGCTTTGCCTACCTGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NWD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NWD1 - NACHT and WD repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007525.3 | Intron | NP_001007526.3 | ||||
NM_001290355.1 | Intron | NP_001277284.1 | ||||
XM_011527929.1 | Intron | XP_011526231.1 | ||||
XM_011527930.2 | Intron | XP_011526232.1 | ||||
XM_011527931.2 | Intron | XP_011526233.1 | ||||
XM_011527934.1 | Intron | XP_011526236.1 | ||||
XM_011527935.1 | Intron | XP_011526237.1 | ||||
XM_017026669.1 | Intron | XP_016882158.1 |