Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAGAAAATGCAAAAGCAGGAAAT[C/G]AACATTTTTCATTGCCTGTTAACCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605529 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UPF2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UPF2 - UPF2 regulator of nonsense transcripts homolog (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015542.3 | Intron | NP_056357.1 | ||||
NM_080599.2 | Intron | NP_542166.1 | ||||
XM_011519449.2 | Intron | XP_011517751.1 | ||||
XM_017016034.1 | Intron | XP_016871523.1 |
Set Membership: |
HapMap |