Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTTTTTCAGTACTTTGTAACAT[C/T]AGTCAAAATAGAATGAGCCAACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605405 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP6NL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP6NL - USP6 N-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080491.3 | 6253 | UTR 3 | NP_001073960.1 | |||
NM_014688.3 | 6253 | UTR 3 | NP_055503.1 | |||
XM_006717542.3 | 6253 | UTR 3 | XP_006717605.1 | |||
XM_011519762.2 | 6253 | UTR 3 | XP_011518064.1 | |||
XM_011519763.2 | 6253 | UTR 3 | XP_011518065.1 | |||
XM_017016971.1 | 6253 | UTR 3 | XP_016872460.1 | |||
XM_017016972.1 | 6253 | UTR 3 | XP_016872461.1 | |||
XM_017016973.1 | 6253 | UTR 3 | XP_016872462.1 |