Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATCCAAAGGCTAAGTACCTAGA[A/G]TGCTTCAAACAGCAAAACCTGATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600835 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf142 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C10orf142 - chromosome 10 open reading frame 142 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CXCL12 - C-X-C motif chemokine ligand 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000609.6 | Intron | NP_000600.1 | ||||
NM_001033886.2 | Intron | NP_001029058.1 | ||||
NM_001178134.1 | Intron | NP_001171605.1 | ||||
NM_001277990.1 | Intron | NP_001264919.1 | ||||
NM_199168.3 | Intron | NP_954637.1 |