Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCATGACATCTCGGAGTTTGAT[G/T]GCTCTTCCTTCCACTTGGCAAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604058 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS3ST3B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HS3ST3B1 - heparan sulfate-glucosamine 3-sulfotransferase 3B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006041.2 | Intron | NP_006032.1 | ||||
XM_017025479.1 | Intron | XP_016880968.1 |
MGC12916 - uncharacterized protein MGC12916 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |