Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCATAAAATTAGGTGCAAAAAT[C/T]GTAGACCTACAAGTGATGTCTTTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604210 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRB1 - crumbs 1, cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193640.1 | Intron | NP_001180569.1 | ||||
NM_001257965.1 | Intron | NP_001244894.1 | ||||
NM_001257966.1 | Intron | NP_001244895.1 | ||||
NM_201253.2 | Intron | NP_957705.1 | ||||
XM_011509365.2 | Intron | XP_011507667.1 | ||||
XM_011509367.1 | Intron | XP_011507669.1 | ||||
XM_011509369.2 | Intron | XP_011507671.1 | ||||
XM_017000851.1 | Intron | XP_016856340.1 | ||||
XM_017000852.1 | Intron | XP_016856341.1 |
ZBTB41 - zinc finger and BTB domain containing 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |