Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTAGTTTCAGTCCTACTCACC[C/T]GAGTTTGCAGTGCTTCTTTGTACCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613326 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPYS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DPYS - dihydropyrimidinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001385.2 | Intron | NP_001376.1 | ||||
XM_005250818.3 | Intron | XP_005250875.1 | ||||
XM_006716518.3 | Intron | XP_006716581.1 | ||||
XM_011516903.2 | Intron | XP_011515205.1 | ||||
XM_017013167.1 | Intron | XP_016868656.1 |
Set Membership: |
HapMap |