Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAAATCGTGATCCTGAAGCCCTA[A/C]ATGATCCCCAATCAAAAAGCAGTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611350 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
INTS7 - integrator complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199809.1 | 4341 | UTR 3 | NP_001186738.1 | |||
NM_001199811.1 | 4341 | UTR 3 | NP_001186740.1 | |||
NM_001199812.1 | 4341 | UTR 3 | NP_001186741.1 | |||
NM_015434.3 | 4341 | UTR 3 | NP_056249.1 | |||
XM_011509396.2 | 4341 | UTR 3 | XP_011507698.1 | |||
XM_017000962.1 | 4341 | Intron | XP_016856451.1 |