Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGGCTAGTTTGCTGGAATGCT[A/G]ACGAATCCAAAGACCCACAGGACTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609527 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAPGEF5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RAPGEF5 - Rap guanine nucleotide exchange factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012294.3 | 7277 | UTR 3 | NP_036426.3 | |||
XM_005249914.2 | 7277 | UTR 3 | XP_005249971.1 | |||
XM_005249916.1 | 7277 | UTR 3 | XP_005249973.1 | |||
XM_011515652.2 | 7277 | UTR 3 | XP_011513954.1 | |||
XM_017012837.1 | 7277 | UTR 3 | XP_016868326.1 |
Set Membership: |
HapMap |