Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGAAAGTGGATGAAAGAAGCT[A/C]TGTCTCCCTGGGACAGATCAGTCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608798 MIM: 606959 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DFNA5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DFNA5 - DFNA5, deafness associated tumor suppressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127453.1 | Intron | NP_001120925.1 | ||||
NM_001127454.1 | Intron | NP_001120926.1 | ||||
NM_004403.2 | Intron | NP_004394.1 | ||||
XM_017011802.1 | Intron | XP_016867291.1 |
MPP6 - membrane palmitoylated protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |