Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGACACAGACCAAATTCAAAGGG[C/T]GCAAAAGGGTATACAGCTTCAAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606732 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OSBPL3 - oxysterol binding protein like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015550.3 | Intron | NP_056365.1 | ||||
NM_145320.2 | Intron | NP_663160.1 | ||||
NM_145321.2 | Intron | NP_663161.1 | ||||
NM_145322.2 | Intron | NP_663162.1 | ||||
XM_005249698.3 | Intron | XP_005249755.1 | ||||
XM_006715681.3 | Intron | XP_006715744.1 | ||||
XM_006715682.3 | Intron | XP_006715745.1 | ||||
XM_006715683.3 | Intron | XP_006715746.1 | ||||
XM_006715684.3 | Intron | XP_006715747.1 | ||||
XM_011515258.2 | Intron | XP_011513560.1 | ||||
XM_011515259.2 | Intron | XP_011513561.1 | ||||
XM_017011948.1 | Intron | XP_016867437.1 | ||||
XM_017011949.1 | Intron | XP_016867438.1 |