Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATCTTCAGTTTCTTTGGACAAA[A/G]GCATACTACTATCTTCTTTTTTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605792 | ||||||||||||||||||||
Literature Links: |
SEPT4-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEPT4-AS1 - SEPT4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX14 - testis expressed 14, intercellular bridge forming factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201457.1 | 4362 | Missense Mutation | CCT,CTT | P,L 1387 | NP_001188386.1 | |
NM_031272.4 | 4362 | Missense Mutation | CCT,CTT | P,L 1341 | NP_112562.3 | |
NM_198393.3 | 4362 | Missense Mutation | CCT,CTT | P,L 1381 | NP_938207.2 | |
XM_011525029.2 | 4362 | Missense Mutation | CCT,CTT | P,L 1422 | XP_011523331.1 | |
XM_017024860.1 | 4362 | Missense Mutation | CCT,CTT | P,L 1200 | XP_016880349.1 | |
XM_017024861.1 | 4362 | Missense Mutation | CCT,CTT | P,L 989 | XP_016880350.1 |