Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATTTTAAATTATACTAGTTGAA[A/C]CTTTATACTACAATGTATCCTGGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35F5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC35F5 - solute carrier family 35 member F5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005263799.3 | 9359 | UTR 3 | XP_005263856.1 | |||
XM_011511921.2 | 9359 | UTR 3 | XP_011510223.1 | |||
XM_011511922.2 | 9359 | UTR 3 | XP_011510224.1 | |||
XM_011511923.2 | 9359 | UTR 3 | XP_011510225.1 | |||
XM_017005027.1 | 9359 | Intron | XP_016860516.1 | |||
XM_017005028.1 | 9359 | UTR 3 | XP_016860517.1 |
Set Membership: |
HapMap |