Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGACTTGGCTACAGCCCTTGCTA[C/T]GTTTGAGATGTTCAGTATTTGTTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607815 MIM: 602233 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKS1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APAF1 - apoptotic peptidase activating factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160.2 | 8014 | Intron | NP_001151.1 | |||
NM_013229.2 | 8014 | Intron | NP_037361.1 | |||
NM_181861.1 | 8014 | Intron | NP_863651.1 | |||
NM_181868.1 | 8014 | Intron | NP_863658.1 | |||
NM_181869.1 | 8014 | Intron | NP_863659.1 | |||
XM_017019250.1 | 8014 | Intron | XP_016874739.1 |
Set Membership: |
HapMap |