Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGAATTCCAGGGTGGATATCTC[A/G]CTAAGGAACACAAGTATCAGAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608561 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STAB2 - stabilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017564.9 | Intron | NP_060034.9 | ||||
XM_011538537.2 | Intron | XP_011536839.1 | ||||
XM_011538538.2 | Intron | XP_011536840.1 | ||||
XM_011538539.2 | Intron | XP_011536841.1 | ||||
XM_011538541.2 | Intron | XP_011536843.1 | ||||
XM_011538542.2 | Intron | XP_011536844.1 | ||||
XM_017019585.1 | Intron | XP_016875074.1 |
Set Membership: |
HapMap |