Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGAGCAGGGAATATAACTGCAA[C/G]AATATTGAAGAAGTTATTCAATGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608564 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GIT2 - GIT ArfGAP 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139201.2 | Intron | NP_631940.2 | ||||
XM_005253997.3 | Intron | XP_005254054.1 | ||||
XM_005254000.3 | Intron | XP_005254057.1 | ||||
XM_006719707.3 | Intron | XP_006719770.1 | ||||
XM_006719708.3 | Intron | XP_006719771.1 | ||||
XM_006719709.3 | Intron | XP_006719772.1 | ||||
XM_006719712.3 | Intron | XP_006719775.1 | ||||
XM_006719713.3 | Intron | XP_006719776.1 | ||||
XM_006719714.3 | Intron | XP_006719777.1 | ||||
XM_017020258.1 | Intron | XP_016875747.1 | ||||
XM_017020259.1 | Intron | XP_016875748.1 | ||||
XM_017020260.1 | Intron | XP_016875749.1 | ||||
XM_017020261.1 | Intron | XP_016875750.1 |