Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTTTCCGGTGCGCCCTGGGCGG[C/T]GTAGATCCGAAGGTGCTCGCACCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985657 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC107985657 - uncharacterized LOC107985657 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017030004.1 | 1591 | Missense Mutation | ACC,GCC | T,A 39 | XP_016885493.1 |
TCEANC - transcription elongation factor A N-terminal and central domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297563.1 | 1591 | Intron | NP_001284492.1 | |||
NM_001297564.1 | 1591 | Intron | NP_001284493.1 | |||
NM_152634.3 | 1591 | UTR 5 | NP_689847.2 | |||
XM_017029316.1 | 1591 | Intron | XP_016884805.1 | |||
XM_017029317.1 | 1591 | Intron | XP_016884806.1 | |||
XM_017029318.1 | 1591 | Intron | XP_016884807.1 | |||
XM_017029319.1 | 1591 | UTR 5 | XP_016884808.1 |
Set Membership: |
HapMap |