Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGTATAAAACTCTAAACCTGGA[G/T]TCTTCTGATATAATAAATTCTTTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300457 MIM: 300227 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NHS PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NHS - NHS actin remodeling regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCML1 - sex comb on midleg-like 1 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037535.2 | Intron | NP_001032624.1 | ||||
NM_001037536.2 | Intron | NP_001032625.1 | ||||
NM_001037540.2 | Intron | NP_001032629.1 | ||||
NM_006746.5 | Intron | NP_006737.2 | ||||
XM_005274578.2 | Intron | XP_005274635.1 | ||||
XM_005274579.2 | Intron | XP_005274636.1 | ||||
XM_006724508.3 | Intron | XP_006724571.1 | ||||
XM_006724509.3 | Intron | XP_006724572.1 | ||||
XM_011545564.2 | Intron | XP_011543866.1 | ||||
XM_017029723.1 | Intron | XP_016885212.1 |
Set Membership: |
HapMap |