Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTAGCAGAGGGTGGAATTACCA[A/G]GAGAAAGAAAACAGTCTCCTCCCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614681 MIM: 147582 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HYKK PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HYKK - hydroxylysine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013619.3 | Intron | NP_001013641.2 | ||||
NM_001083612.1 | Intron | NP_001077081.1 | ||||
XM_011521231.2 | Intron | XP_011519533.1 | ||||
XM_017021922.1 | Intron | XP_016877411.1 |
IREB2 - iron responsive element binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |