Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTACCCAGGCTCTCCAAGGCTGAG[A/G]AACATGGATGGCACAGCTCTCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601465 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGUOK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DGUOK - deoxyguanosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318859.1 | Intron | NP_001305788.1 | ||||
NM_001318860.1 | Intron | NP_001305789.1 | ||||
NM_001318861.1 | Intron | NP_001305790.1 | ||||
NM_001318862.1 | Intron | NP_001305791.1 | ||||
NM_001318863.1 | Intron | NP_001305792.1 | ||||
NM_080916.2 | Intron | NP_550438.1 | ||||
NM_080918.2 | Intron | NP_550440.1 | ||||
XM_011532647.2 | Intron | XP_011530949.1 |
Set Membership: |
HapMap |