Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGTTATGCCTCCATGTTACCAT[A/C]CAATAACAAGGACAGAGATGCCTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600021 | ||||||||||||||||||||
Literature Links: |
MXD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MXD1 - MAX dimerization protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202513.1 | 397 | Missense Mutation | TAC,TCC | Y,S 36 | NP_001189442.1 | |
NM_001202514.1 | 397 | Missense Mutation | TAC,TCC | Y,S 36 | NP_001189443.1 | |
NM_002357.3 | 397 | Missense Mutation | TAC,TCC | Y,S 36 | NP_002348.1 |