Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGCAGCTACGGCCAAGGCTACC[A/C]TACGCCCTCCTCTTCCTACTTTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609922 MIM: 600036 | ||||||||||||||||||||
Literature Links: |
EHBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EHBP1 - EH domain binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100132215 - uncharacterized LOC100132215 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTX1 - orthodenticle homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199770.1 | 983 | Missense Mutation | CAT,CCT | H,P 236 | NP_001186699.1 | |
NM_014562.3 | 983 | Missense Mutation | CAT,CCT | H,P 236 | NP_055377.1 |