Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTCAGTCATGCAACGAACCCTC[A/G]ACACTTGGAAGGTGACAGAGGGTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612396 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALLC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ALLC - allantoicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018436.3 | Intron | NP_060906.3 | ||||
XM_011510369.2 | Intron | XP_011508671.1 | ||||
XM_011510370.2 | Intron | XP_011508672.1 | ||||
XM_017004495.1 | Intron | XP_016859984.1 | ||||
XM_017004496.1 | Intron | XP_016859985.1 | ||||
XM_017004497.1 | Intron | XP_016859986.1 | ||||
XM_017004498.1 | Intron | XP_016859987.1 |
Set Membership: |
HapMap |