Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTACTTACTGGGAAAGGTGATAAA[A/G]GCCTGGCCCCTCATTCGTCCAGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RBM41 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM41 - RNA binding motif protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171080.1 | 1185 | Silent Mutation | GCC,GCT | A,A 353 | NP_001164551.1 | |
NM_001324242.1 | 1185 | Silent Mutation | GCC,GCT | A,A 377 | NP_001311171.1 | |
NM_001324243.1 | 1185 | Silent Mutation | GCC,GCT | A,A 412 | NP_001311172.1 | |
NM_001324244.1 | 1185 | Silent Mutation | GCC,GCT | A,A 377 | NP_001311173.1 | |
NM_018301.4 | 1185 | Silent Mutation | GCC,GCT | A,A 353 | NP_060771.3 | |
XM_011530980.2 | 1185 | Silent Mutation | GCC,GCT | A,A 412 | XP_011529282.1 | |
XM_011530981.2 | 1185 | Silent Mutation | GCC,GCT | A,A 388 | XP_011529283.1 |