Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCAGATATGGCACCCCGATCAG[A/T]CCCCAGTCCGACAAGGTATGATGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604863 | ||||||||||||||||||||
Literature Links: |
LRAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRAT - lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301645.1 | 584 | Silent Mutation | AGA,AGT | R,S 175 | NP_001288574.1 | |
NM_004744.4 | 584 | Silent Mutation | AGA,AGT | R,S 175 | NP_004735.2 |