Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGATTTTATTATTTTGAGAATTC[A/G]TGGCATCTTCCATGAGAGTTTATAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610514 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JADE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
JADE1 - jade family PHD finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287437.1 | Intron | NP_001274366.1 | ||||
NM_001287439.1 | Intron | NP_001274368.1 | ||||
NM_001287440.1 | Intron | NP_001274369.1 | ||||
NM_001287441.1 | Intron | NP_001274370.1 | ||||
NM_001287442.1 | Intron | NP_001274371.1 | ||||
NM_001287443.1 | Intron | NP_001274372.1 | ||||
NM_024900.4 | Intron | NP_079176.2 | ||||
NM_199320.3 | Intron | NP_955352.1 | ||||
XM_005263232.1 | Intron | XP_005263289.1 | ||||
XM_017008626.1 | Intron | XP_016864115.1 | ||||
XM_017008627.1 | Intron | XP_016864116.1 |
Set Membership: |
HapMap |