Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTGGCAGAGCTGAGCCTCCCTC[G/T]GGTCTTCTGGTGGCAAGATGCCAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300694 MIM: 300353 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR223 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR223 - microRNA 223 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VSIG4 - V-set and immunoglobulin domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100431.1 | 1664 | UTR 3 | NP_001093901.1 | |||
NM_001184830.1 | 1664 | UTR 3 | NP_001171759.1 | |||
NM_001184831.1 | 1664 | UTR 3 | NP_001171760.1 | |||
NM_001257403.1 | 1664 | UTR 3 | NP_001244332.1 | |||
NM_007268.2 | 1664 | UTR 3 | NP_009199.1 | |||
XM_017029251.1 | 1664 | UTR 3 | XP_016884740.1 |
Set Membership: |
HapMap |