Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGGGCCAACGTAATTGCAACAG[-/C]CCTTGCCAAGATCCCACAAAAGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600070 | ||||||||||||||||||||||||||||||||
Literature Links: |
UGT2B10 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
UGT2B10 - UDP glucuronosyltransferase family 2 member B10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001075.5 | Intron | NP_001066.1 | ||||
NM_001144767.2 | Intron | NP_001138239.1 | ||||
NM_001290091.1 | Intron | NP_001277020.1 |