Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACAGCTTCTATTAAAAATGCCG[C/T]CATTATTTAAATGATGTTGTTGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SDAD1 - SDA1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288983.1 | Intron | NP_001275912.1 | ||||
NM_001288984.1 | Intron | NP_001275913.1 | ||||
NM_018115.3 | Intron | NP_060585.2 | ||||
XM_005263102.4 | Intron | XP_005263159.1 | ||||
XM_005263105.4 | Intron | XP_005263162.1 | ||||
XM_017008354.1 | Intron | XP_016863843.1 | ||||
XM_017008355.1 | Intron | XP_016863844.1 |
Set Membership: |
HapMap |