Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGTAAACTTCTCAAAAAACCAA[G/T]CCAAGAAAGAATTGTTTTCTTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603086 MIM: 607607 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ART3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ART3 - ADP-ribosyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP54 - nucleoporin 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278603.1 | 1755 | UTR 3 | NP_001265532.1 | |||
NM_017426.3 | 1755 | UTR 3 | NP_059122.2 | |||
XM_006714237.1 | 1755 | UTR 3 | XP_006714300.1 | |||
XM_011532033.2 | 1755 | UTR 3 | XP_011530335.1 | |||
XM_011532034.2 | 1755 | UTR 3 | XP_011530336.1 | |||
XM_017008294.1 | 1755 | UTR 3 | XP_016863783.1 | |||
XM_017008295.1 | 1755 | UTR 3 | XP_016863784.1 |