Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATACATGCTCTAGTTTTTGGAAC[A/C]ATGTCATTAAGTTTTCAAATACATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614532 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASGEF1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RASGEF1B - RasGEF domain family member 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300735.1 | 2851 | UTR 3 | NP_001287664.1 | |||
NM_001300736.1 | 2851 | UTR 3 | NP_001287665.1 | |||
NM_152545.2 | 2851 | UTR 3 | NP_689758.1 | |||
XM_017007812.1 | 2851 | UTR 3 | XP_016863301.1 | |||
XM_017007813.1 | 2851 | UTR 3 | XP_016863302.1 |
Set Membership: |
HapMap |