Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGGCAGTAGTCGGGTGGCCAA[G/T]ACCAACTCCTTACGGAGGCGGAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614799 MIM: 609977 | ||||||||||||||||||||
Literature Links: |
C1orf109 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDCA8 - cell division cycle associated 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256875.1 | 229 | Missense Mutation | AAG,AAT | K,N 12 | NP_001243804.1 | |
NM_018101.3 | 229 | Missense Mutation | AAG,AAT | K,N 12 | NP_060571.1 |