Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCAGAATTCATGAACCTATGCT[C/T]TTTCCTTTATACCACAAAACTGGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147572 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFI6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IFI6 - interferon alpha inducible protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002038.3 | Intron | NP_002029.3 | ||||
NM_022872.2 | Intron | NP_075010.1 | ||||
NM_022873.2 | Intron | NP_075011.1 |