Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAGCCTTGGGCCTTTCTGAAGA[C/G]ATAGCGCCTAGTGGGAGCCAGGGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611499 MIM: 608838 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUSB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GUSB - glucuronidase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000181.3 | Intron | NP_000172.2 | ||||
NM_001284290.1 | Intron | NP_001271219.1 | ||||
NM_001293104.1 | Intron | NP_001280033.1 | ||||
NM_001293105.1 | Intron | NP_001280034.1 | ||||
XM_005250297.3 | Intron | XP_005250354.1 | ||||
XM_011516113.1 | Intron | XP_011514415.1 | ||||
XM_011516114.1 | Intron | XP_011514416.1 | ||||
XM_017012091.1 | Intron | XP_016867580.1 | ||||
XM_017012092.1 | Intron | XP_016867581.1 | ||||
XM_017012093.1 | Intron | XP_016867582.1 |
VKORC1L1 - vitamin K epoxide reductase complex subunit 1 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |