Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGACAGGGTTTGAATCTGTGGAG[A/G]CATACTGTCGGTGAACCCCTCTGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615712 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTULIN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OTULIN - OTU deubiquitinase with linear linkage specificity | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138348.4 | Intron | NP_612357.4 | ||||
XM_011514151.2 | Intron | XP_011512453.1 | ||||
XM_011514152.2 | Intron | XP_011512454.1 | ||||
XM_011514154.2 | Intron | XP_011512456.1 | ||||
XM_017010015.1 | Intron | XP_016865504.1 |