Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147460 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOD2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SOD2 - superoxide dismutase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000636.3 | 1465 | UTR 3 | NP_000627.2 | |||
NM_001024465.2 | 1465 | Intron | NP_001019636.1 | |||
NM_001024466.2 | 1465 | Intron | NP_001019637.1 | |||
NM_001322814.1 | 1465 | UTR 3 | NP_001309743.1 | |||
NM_001322815.1 | 1465 | UTR 3 | NP_001309744.1 | |||
NM_001322816.1 | 1465 | Intron | NP_001309745.1 | |||
NM_001322817.1 | 1465 | Intron | NP_001309746.1 | |||
NM_001322819.1 | 1465 | UTR 3 | NP_001309748.1 | |||
NM_001322820.1 | 1465 | UTR 3 | NP_001309749.1 |
Set Membership: |
HapMap |